missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Contec™ Polynit™ Wipes
Clean surfaces of products and process tools. The Contec™ Polynit™ Wipes are a durable option for ISO Class 5 environments.
Manufacturer: Contec™ PN99B
Description
- For general-purpose cleaning in ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
- Ideal for wiping exterior surfaces of products and cleaning interior surfaces of process tools or other equipment
Specifications
9 in. | |
ISO Class 5 | |
9 x 9 in. (23 x 23 cm) | |
9 in. | |
Polyester | |
9 in. | |
9 in. |
23 cm | |
White | |
Cleanroom | |
23 cm | |
Polynit™ Wipes | |
300 wipes/bag, Bulk | |
Non-sterile |