Contec™ Amplitude™ Kappa™ Wipes
Provide an exceptional level of cleanliness, sorbency and softness. Contec™ Amplitude™ Kappa™ Wipes are used for wiping delicate surfaces and sensitive components, as well as for other applications. A special hydroentangled process washes the Amplitude Kappa fabric, which is made of 55% polyester and 45% lyocell.
Manufacturer:  Contec™ AMLP0001
 View more versions of this product
Catalog No. 19-130-5919
Description
Description
  • Made of hydroentangled lyocell (45%)/polyester(55%) nonwoven for exceptional cleanliness
  • Highly sorbent with water and solvents
  • Soft, low-particulate cleanroom fabric
  • Double packed to preserve product integrity
Specifications
Specifications
ISO Class 5
9 in.
9 in.
Cleanroom
23 cm
Amplitude™ Kappa cleanroom wipes
9 x 9 in. (23 x 23 cm)
9 in.
23 cm
9 in.
Lyocell, Polyester
Documents