Contec™ Amplitude™ Epsilon™ Cleanroom Wipes
Equip workers with the tools to keep cleanrooms clean. The Amplitude™ Epsilon™ Cleanroom Wipes, are a durable solution for ISO Class 6 applications.
Manufacturer:  Contec™ AMEP0002
 View more versions of this product
Catalog No. 19-130-5930
Description
Description
  • Polyester/cellulose hydroentangled nonwoven wipes are strong and durable and provide improved cleanliness
  • Heavyweight, with high sorbency
  • Dye-fast blue color for differentiation between areas of use
  • Excellent all-purpose cleanroom wipes
  • Lot-numbered for traceability
  • Made of hydroentangled premium polyester/cellulose for improved cleanliness
  • Excellent for wiping and absorbing spills and in general purpose cleaning in biotech, pharmaceutical, semiconductor, electronics, medical device, and printed circuit board industries
  • Double packed to preserve product integrity
Recommendation: Test colorfastness with each process before use.
Specifications
Specifications
ISO Class 6
12 x 12 in. (30.5 x 30.5 cm)
12 in.
30.5 cm
12 in.
Cellulose, Polyester
Blue
12 in.
12 in.
Cleanroom
30.5 cm
Amplitude™ Epsilon cleanroom wipes
Documents