Contec™ Amplitude™ Epsilon™ Cleanroom Wipes
Equip workers with the tools to keep cleanrooms clean. The Amplitude™ Epsilon™ Cleanroom Wipes, are a durable solution for ISO Class 6 applications.
Supplier:  Contec™ AMEP0001
 View more versions of this product
Catalog No. 19-130-5929
Description
Description
  • Polyester/cellulose hydroentangled nonwoven wipes are strong and durable and provide improved cleanliness
  • Heavyweight, with high sorbency
  • Dye-fast blue color for differentiation between areas of use
  • Excellent all-purpose cleanroom wipes
  • Lot-numbered for traceability
  • Made of hydroentangled premium polyester/cellulose for improved cleanliness
  • Excellent for wiping and absorbing spills and in general purpose cleaning in biotech, pharmaceutical, semiconductor, electronics, medical device, and printed circuit board industries
  • Double packed to preserve product integrity
Recommendation: Test colorfastness with each process before use.
Specifications
Specifications
Amplitude™ Epsilon Cleanroom Wipes
23 cm
23 cm
ISO Class 6
9 x 9 in. (22.86 x 22.86 cm)
9 in.
9 in.
Polyester Cellulose
Blue
Cleanroom