Contec™ ABRATEC™ Diamond Grit Abrasives with Quiltec™ Backing
Remediate stainless steel, remove disinfectant residue and minimize rouging due to residues and soils. The Contec™ ABRATEC™ Diamond Grit Abrasives with Quiltec™; Backing are designed to work with non-ketone based solvents. Compatible with autoclave and gamma irradiation processing to help maintain the integrity of sterile environments.
$537.86 - $556.87
Specifications
For Use With (Application) Removal of residues from stainless steel surfaces
Material Quiltec™
Size 8 x 10cm (3 x 4 in.)
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Grit Rating Price Quantity  
Catalog Number Mfr. No. Grit Rating Price Quantity  
19-033-412 Contec™
ATQT1350
1350 Case of 10 Each for $543.28
19-033-410 Contec™
ATQT0280
280 Case of 10 Each for $556.87
19-033-411 Contec™
ATQT0360
360 Case of 10 Each for $537.86
Specifications
Description
  •  ABRATEC abrasive cloths are made with various levels of abrasive grits backed with Contec’s unique Quiltec fabric
  • Quiltec fabric is significantly more durable and abrasion resistant than similar foam-backed abrasives
  • The abrasion resistance and the Contec sealed border technology allow the Quiltec side to remove sanding debris without contributing additional particles to the remediation process
  • ABRATEC abrasive cloths are designed to work with solvents, including the IPA/water blends typically found in sterile processing environments
  • ABRATEC cloths are also available irradiated with a dose of 25-50 kGy, which helps maintain the integrity of sterile environments
  • ABRATEC cloths are faster, cleaner, have lower particulation, and are safer than similar foam-backed abrasive products
Description & Specifications
Specifications
Removal of residues from stainless steel surfaces
Quiltec™
8 x 10cm (3 x 4 in.)