Please login to your online account to display your discounted pricing

Connexin 43 Mouse anti-Mouse, eFluor 660, Clone: Connexin43 (1A), eBioscience™

Mouse Monoclonal Antibody

$172.55 - $433.55

Specifications

Antigen Connexin 43
Clone Connexin43 (1A)
Host Species Mouse
Species Reactivity Human, Mouse, Rat
Applications Immunocytochemistry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501124362

 
affymetrix
50475980
25μg Each for $172.55

501124363

 
affymetrix
50475982
100μg Each for $433.55
Description & Specifications

Specifications

Antigen Connexin 43
Clone Connexin43 (1A)
Host Species Mouse
Species Reactivity Human, Mouse, Rat
Applications Immunocytochemistry
Applications Immunohistochemistry (Frozen)
Applications Microscopy
Regulatory Status RUO
Conjugate eFluor 660
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The Connexin43 monoclonal antibody recognizes mouse Connexin 43. Connexins are a family of integral membrane proteins that form gap junction channels, which function to facilitate direct cell-cell communication. Gap junctions are comprised of two hemichannels, or connexons, which are made up of six connexin proteins. Over 20 connexin proteins have been identified and each combine to form either homomeric or heteromeric channels with different functional properties. Connexin 43 is expressed in many tissues, including the central nervous system, heart, and bone. Connexin 43 is the most abundant connexin found in the myocardium and is also the primary connexin expressed in astrocytes. Mutations in connexin 43 are associated with ocluodentodigital dysplasia, a rare developmental disorder characterized by craniofacial and limb abnormalities. Connexin 43 may also be a promising therapeutic target in the treatment of malignant melanomas and mammary gland tumors.

The monoclonal antibody Connexin43 also recognizes human and rat connexin 43.