missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Connecting Microcentrifuge Tube Racks

Maximize storage and save benchtop space with microcentrifuge tube racks that are stackable when loaded or empty

$44.60 - $44.60

Specifications

Dimensions (L x W x H) 10.62 x 1.75 x 2.5 in. (26.8 x 4.8 x 6.4cm)
Material Polycarbonate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Holds Price Quantity  
Catalog Number Mfr. No. Holds Price Quantity  
S90132A Fisher Science Education
F18908-0015
1.5 to 2mL Tubes Each for $44.60
Specifications

Description

Locking clips firmly connect multiple racks together to form larger arrays that can be easily transported.

  • Securely hold 24 microcentrifuge tubes, either 0.5mL or 1.5 to 2.0mL
  • Can be combined when multiple tube sizes are used
  • Durable, lightweight polycarbonate; UV resistant
  • Can be stored at temperatures down to -80°C
  • Steam autoclavable at 121°C (250°F)

Description & Specifications

Specifications

10.62 x 1.75 x 2.5 in. (26.8 x 4.8 x 6.4cm)
Polycarbonate