MilliporeSigma™ Supelco™ Conical Addition Funnel
For charging columns
$180.54 - $561.20
Specifications
For Use With (Application) Charging Columns
Product Type Conical Addition Funnel
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Price Quantity  
111010714
View Documents Promotion Details
MilliporeSigma™ Supelco™
Z2178831EA
300 mL
Each for $180.54
111009216
View Documents Promotion Details
MilliporeSigma™ Supelco™
Z2179131EA
3000 mL
Each for $561.20
Description
Description
  • Capacity is approximately five times the largest column volume
  • Size listed refers to I.D. of tubing above thread and corresponds to M-M column diameter
Specifications
Specifications
Charging Columns
Conical Addition Funnel