Please login to your online account to display your discounted pricing

Congo Red TS, Ricca Chemical

$135.42 - $135.42

Chemical Identifiers

Chemical Name or Material Congo Red
Name Note Test Solution
CAS 7732-18-5
CAS Min % 90.0
CAS Max % 94.0
CAS 64-17-5
CAS Min % 6.54
CAS Max % 6.8
CAS 573-58-0
CAS Min % 0.45
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

2253-16

 
ricca chemical company
RI225316
Poly natural 500mL Each for $135.42

2253-4

 
ricca chemical company
RI22534
Poly natural 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear reddish-orange
Quantity 500mL
CAS 7732-18-5 (Water, Deionized)
Name Note Test Solution
CAS 64-17-5
CAS 573-58-0
Chemical Name or Material Congo Red
CAS Max % 0.47
CAS 67-63-0
CAS Min % 0.35
CAS Max % 0.36
CAS 7732-18-5
CAS Min % 90.0
Packaging Poly natural
CAS Max % 94.0
CAS Min % 6.54
CAS Max % 6.8
CAS Min % 0.45