Please login to your online account to display your discounted pricing

Congo Red, Reagent Grade, LabChem™

$123.23 - $123.23

Chemical Identifiers

Chemical Name or Material Congo Red
CAS 573-58-0
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula C32H22N6O6S2Na2
Formula Weight 696.67
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC133507

 
lab chem
LC133507
10g Each for $123.23
Description & Specifications

Specifications

Formula Weight 696.67
Chemical Name or Material Congo Red
Quantity 10g
EINECS Number 209-358-4
Purity Grade Reagent Grade
Physical Form Powder
Linear Formula C32H22N6O6S2Na2
CAS 573-58-0
Identification Passes Test
Health Hazard 1 Danger
Packaging Amber Glass
CAS Max % 100
Health Hazard 2 GHS H Statement
May cause cancer.
Suspected of damaging fertility or the unborn child.
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
If exposed or concerned: Get medical advice/attention.
Solubility Soluble in water
Recommended Storage Room Temperature
Decomposition Information Nitrogen oxides; Sulfur compounds; Fume; Carbon monoxide; Carbon dioxide
Melting Point 360°C
Color Red
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Wear protective gloves, eye protection.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
If exposed or concerned: Get medical advice/attention.

GHS H Statement
May cause cancer.
Suspected of damaging fertility or the unborn child.

Room Temperature