Please login to your online account to display your discounted pricing

Congo Red Paper, Test Strips, Reagent Grade, LabChem™


Quantity 1 Vial

Manufacturer: lab chem  LC133808

 View more versions of this product

Catalog No. LC133808

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 696.67
Physical Form Paper Strips
Packaging Vial
Chemical Name or Material Congo Red Paper
Quantity 1 Vial
EINECS Number 209-358-4
Name Note Test Strips
Purity Grade Reagent Grade
Linear Formula C32H22N6O6S2Na2
Identification Passes Test
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
May cause cancer.
Suspected of damaging fertility or the unborn child.
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
If exposed or concerned: Get medical advice/attention.
Wear protective gloves, eye protection.
Store locked up.
Dispose of contents to comply with local, state and federal regulations.
Solubility Soluble in water
Recommended Storage Room Temperature
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds
Melting Point 360°C
Color Orange

Specifications

Chemical Name or Material Congo Red Paper
Name Note Test Strips
Purity Grade Reagent Grade
Linear Formula C32H22N6O6S2Na2
Formula Weight 696.67
Accessories

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
If exposed or concerned: Get medical advice/attention.
Wear protective gloves, eye protection.
Store locked up.
Dispose of contents to comply with local, state and federal regulations.

GHS H Statement
May cause cancer.
Suspected of damaging fertility or the unborn child.

Room Temperature