Please login to your online account to display your discounted pricing

Congo Red Paper, Test Strips, Reagent Grade, LabChem™

$18.29 - $18.29

Chemical Identifiers

Chemical Name or Material Congo Red Paper
Name Note Test Strips
Purity Grade Reagent Grade
Linear Formula C32H22N6O6S2Na2
Formula Weight 696.67
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC133808

 
lab chem
LC133808
1 Vial Each for $18.29
Description & Specifications

Specifications

Formula Weight 696.67
Physical Form Paper Strips
Packaging Vial
Chemical Name or Material Congo Red Paper
Quantity 1 Vial
EINECS Number 209-358-4
Name Note Test Strips
Purity Grade Reagent Grade
Linear Formula C32H22N6O6S2Na2
Identification Passes Test
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
May cause cancer.
Suspected of damaging fertility or the unborn child.
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
If exposed or concerned: Get medical advice/attention.
Wear protective gloves, eye protection.
Store locked up.
Dispose of contents to comply with local, state and federal regulations.
Solubility Soluble in water
Recommended Storage Room Temperature
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide; Sulfur compounds
Melting Point 360°C
Color Orange
Safety and Handling

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
If exposed or concerned: Get medical advice/attention.
Wear protective gloves, eye protection.
Store locked up.
Dispose of contents to comply with local, state and federal regulations.

GHS H Statement
May cause cancer.
Suspected of damaging fertility or the unborn child.

Room Temperature