missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing
Conductivity Standard, 445 μS/cm at 25°C (213 ppm TDS as NaCl), Ricca Chemical
Chemical Identifiers
CAS |
7647-14-5 |
Molecular Formula |
ClNa |
Molecular Weight (g/mol) |
58.44 |
MDL Number |
MFCD00003477 |
InChI Key |
FAPWRFPIFSIZLT-UHFFFAOYSA-M |
PubChem CID |
5234 |
ChEBI |
CHEBI:26710 |
IUPAC Name |
sodium chloride |
SMILES |
[Na+].[Cl-] |
View More Specs
Products
2
Description
Specifications
Chemical Identifiers
|
7647-14-5 |
|
58.44 |
|
FAPWRFPIFSIZLT-UHFFFAOYSA-M |
|
CHEBI:26710 |
|
[Na+].[Cl-] |
|
ClNa |
|
MFCD00003477 |
|
5234 |
|
sodium chloride |
Specifications
|
7647-14-5,7732-18-5 |
|
500 mL |
|
213ppm as NaCl |
|
MFCD00003477 |
|
[Na+].[Cl-] |
|
58.44 |
|
CHEBI:26710 |
|
Conductivity/TDS Standard |
|
Natural Poly Bottle |
|
Colorless |
|
ClNa |
|
FAPWRFPIFSIZLT-UHFFFAOYSA-M |
|
sodium chloride |
|
5234 |
|
Liquid |
SDS
missing translation for 'documents'