missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Conductivity Standard, 100,000 μS/cm (100 mS/cm) at 25°C, Ricca Chemical
65,750 ppm TDS as NaCl
Supplier: Ricca Chemical Company RI22491
Specifications
| Conductivity/TDS Standard | |
| Natural Poly Bottle | |
| 93.0,4.85 | |
| Liquid | |
| Approximately 0°C | |
| Conductivity Standard | |
| 65, 750ppm as NaCl |
| 100mS/cm (100, 000μmho/cm) at 25°C | |
| 7647-14-5,7732-18-5 | |
| 97.0,5.05 | |
| Colorless | |
| Conductivity Standard | |
| 4 L |