missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Conductivity Standard, 8974μmho/cm, Certified, LabChem™

$75.60 - $75.60

Chemical Identifiers

CAS 7732-18-5,7447-40-7
CAS Max % 99.5,0.5
Linear Formula KCl
Formula Weight 74.55
Chemical Name or Material Conductivity Standard, 8974μmho/cm
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
LC187871 Lab Chem
LC187871
Poly Bottle 500mL Each for $75.60
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7447-40-7
KCl
Conductivity Standard, 8974μmho/cm
99.5,0.5
74.55
Certified Grade

Specifications

74.55
7732-18-5,7447-40-7
KCl
500mL
Poly Bottle
Liquid
Conductivity Standard, 8974μmho/cm
Passes Test
99.5,0.5
Clear, colorless
1g/mL
Soluble in water
Traceable to NIST
Certified Grade

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature