Please login to your online account to display your discounted pricing

Conductivity Standard, 5000μmho/cm, Certified, LabChem™

$54.60 - $54.60

Chemical Identifiers

Chemical Name or Material Conductivity Standard
CAS 7732-18-5
CAS Max % 99.72
CAS 7447-40-7
CAS Max % 0.28
Name Note 5000μmho/cm
Purity Grade Certified Grade
Linear Formula KCl
Formula Weight 74.55
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC187752

 
lab chem
LC187752
Poly Bottle 1L Each for $54.60
Description & Specifications

Specifications

Formula Weight 74.55
Purity Grade Certified Grade
Traceability to NIST Traceable to NIST
Density 1g/mL
Color Clear, colorless
CAS 7447-40-7
Packaging Poly Bottle
CAS Max % 99.72
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Solubility Soluble in water
Linear Formula KCl
Quantity 1L
Chemical Name or Material Conductivity Standard
Identification Passes Test
Name Note 5000μmho/cm
CAS Max % 0.28
Physical Form Liquid
CAS 7732-18-5
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature