Please login to your online account to display your discounted pricing

Conductivity Standard, 500μmho/cm, LabChem™

$46.14 - $46.14

Chemical Identifiers

Chemical Name or Material Conductivity Standard, 500μmho/cm
CAS 7732-18-5
CAS Max % 99.97
CAS 7447-40-7
CAS Max % 0.03
Purity Grade Certified Grade
Linear Formula KCl
Formula Weight 74.55
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC187602

 
lab chem
LC187602
Poly Bottle 1L Each for $46.14
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 74.55
Solubility Soluble in water
Chemical Name or Material Conductivity Standard, 500μmho/cm
Physical Form Liquid
CAS Max % 0.03
Traceability to NIST Traceable to NIST
Recommended Storage Room Temperature
Color clear, colorless solution
CAS Max % 99.97
Quantity 1L
Packaging Poly Bottle
Linear Formula KCl
CAS 7732-18-5
Identification Passes Test
Density 1g/mL
CAS 7447-40-7
Safety and Handling

Room Temperature