Please login to your online account to display your discounted pricing

Conductivity Standard, 447μmho/cm, LabChem™

$45.60 - $45.60

Chemical Identifiers

Chemical Name or Material Conductivity Standard, 447μmho/cm
CAS 7732-18-5
CAS Max % 99.98
CAS 7447-40-7
CAS Max % 0.02
Purity Grade Certified Grade
Linear Formula KCl
Formula Weight 74.55
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC187551

 
lab chem
LC187551
Poly Bottle 500mL Each for $45.60
Description & Specifications

Specifications

Chemical Name or Material Conductivity Standard, 447μmho/cm
CAS 7732-18-5
CAS Max % 99.98
CAS 7447-40-7
CAS Max % 0.02
Recommended Storage Room Temperature
Purity Grade Certified Grade
Physical Form Liquid
Linear Formula KCl
Formula Weight 74.55
Identification Passes Test
Packaging Poly Bottle
Solubility Soluble in water
Traceability to NIST Traceable to NIST
Density 1g/mL
Color clear, colorless solution
Quantity 500mL
Safety and Handling

Room Temperature