Please login to your online account to display your discounted pricing

Conductivity Standard, 3000μmho/cm, Certified, LabChem™

$73.46 - $73.46

Chemical Identifiers

Chemical Name or Material Conductivity Standard
CAS 7732-18-5
CAS Max % 99.84
CAS 7447-40-7
CAS Max % 0.16
Name Note 3000μmho/cm
Purity Grade Certified Grade
Linear Formula KCl
Formula Weight 74.55
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC187742

 
lab chem
LC187742
Poly Bottle 1L Each for $73.46
Description & Specifications

Specifications

Chemical Name or Material Conductivity Standard
Name Note 3000μmho/cm
CAS 7732-18-5
CAS Max % 99.84
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 7447-40-7
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 0.16
Recommended Storage Room Temperature
Purity Grade Certified Grade
Physical Form Liquid
Linear Formula KCl
Formula Weight 74.55
Identification Passes Test
Packaging Poly Bottle
Solubility Soluble in water
Traceability to NIST Traceable to NIST
Density 1g/mL
Color Clear, colorless
Quantity 1L
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature