Please login to your online account to display your discounted pricing

Conductivity Standard, 1500μmho/cm, LabChem™

$67.03 - $67.03

Chemical Identifiers

Chemical Name or Material Conductivity Standard, 1500μmho/cm
CAS 7732-18-5
CAS Max % 99.92
CAS 7447-40-7
CAS Max % 0.08
Purity Grade Certified Grade
Linear Formula KCl
Formula Weight 74.55
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC187861

 
lab chem
LC187861
Poly Bottle 500mL Each for $67.03
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 74.55
Chemical Name or Material Conductivity Standard, 1500μmho/cm
Color clear, colorless solution
Quantity 500mL
Physical Form Liquid
Specific Gravity 1g/mL
Linear Formula KCl
CAS 7732-18-5
Decomposition Information Hydrogen chloride; Potassium oxide
Conductivity 1500umho/cm ± 20
Identification Passes Test
CAS Max % 99.92
CAS 7447-40-7
Packaging Poly Bottle
Solubility Soluble in water
Traceability to NIST Traceable to NIST
CAS Max % 0.08
Recommended Storage Room Temperature
Density 1g/mL
Safety and Handling

Room Temperature