Please login to your online account to display your discounted pricing

Conductivity Standard, 15,000μmho/cm, Certified, LabChem™

$68.91 - $68.91

Chemical Identifiers

Chemical Name or Material Conductivity Standard
CAS 7732-18-5
CAS Max % 99.14
CAS 7447-40-7
CAS Max % 0.86
Name Note 15,000μmho/cm
Purity Grade Certified Grade
Linear Formula KCl
Formula Weight 74.55
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC187891

 
lab chem
LC187891
Poly Bottle 500mL Each for $68.91
Description & Specifications

Specifications

Formula Weight 74.55
Purity Grade Certified Grade
CAS 7447-40-7
Health Hazard 2 GHS H Statement
Product is not hazardous.
Quantity 500mL
Packaging Poly Bottle
Chemical Name or Material Conductivity Standard
Physical Form Liquid
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
CAS Max % 0.86
CAS Max % 99.14
Traceability to NIST Traceable to NIST
Color Clear, colorless
Identification Passes Test
Density 1g/mL
Linear Formula KCl
CAS 7732-18-5
Recommended Storage Room Temperature
Solubility Soluble in water
Name Note 15,000μmho/cm
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.

Room Temperature