Conductivity Standard, 1409μmho/cm, Certified, LabChem™
Manufacturer:  LabChem LC187806
 View more versions of this product
Catalog No. LC187806
Specifications
Specifications
Conductivity Standard
7447-40-7,7732-18-5
KCl
Liquid
Certified
Soluble in water
1g/mL
10L
1409μmho/cm
99.93,0.07
74.55
Passes Test
Cubitainer™
Traceable to NIST
Colorless