Please login to your online account to display your discounted pricing

Conductivity Standard, 12,856μmho/cm, Certified, LabChem™

$68.60 - $68.60

Chemical Identifiers

Chemical Name or Material Conductivity Standard
CAS 7732-18-5
CAS Max % 99.25
CAS 7447-40-7
CAS Max % 0.75
Name Note 12,856μmho/cm
Purity Grade Certified Grade
Linear Formula KCl
Formula Weight 74.55
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC187792

 
lab chem
LC187792
Poly Bottle 1L Each for $68.60
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 74.55
Chemical Name or Material Conductivity Standard
Name Note 12,856μmho/cm
Specific Gravity 1g/mL
Linear Formula KCl
CAS 7732-18-5
Decomposition Information Hydrogen chloride; Potassium oxide
Conductivity 12856umho/cm ±100
CAS 7447-40-7
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
Traceability to NIST Traceable to NIST
CAS Max % 0.75
Density 1g/mL
Color Clear, colorless
Quantity 1L
Physical Form Liquid
Identification Passes Test
CAS Max % 99.25
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Solubility Soluble in water
Recommended Storage Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature