Please login to your online account to display your discounted pricing

Conductivity Standard, 10,000μmho/cm, Certified, LabChem™

$73.46 - $73.46

Chemical Identifiers

Chemical Name or Material Conductivity Standard
CAS 7732-18-5
CAS Max % 99.43
CAS 7447-40-7
CAS Max % 0.57
Name Note 10,000μmho/cm
Purity Grade Certified Grade
Linear Formula KCl
Formula Weight 74.55
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC187772

 
lab chem
LC187772
Poly Bottle 1L Each for $73.46
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 74.55
Chemical Name or Material Conductivity Standard
Color Clear, colorless
Quantity 1L
Name Note 10,000μmho/cm
Physical Form Liquid
Specific Gravity 1g/mL
Linear Formula KCl
CAS 7732-18-5
Decomposition Information Hydrogen chloride; Potassium oxide
Conductivity 10000umho/cm ±100
Identification Passes Test
CAS Max % 99.43
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 7447-40-7
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
Solubility Soluble in water
Traceability to NIST Traceable to NIST
CAS Max % 0.57
Recommended Storage Room Temperature
Density 1g/mL
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature