Please login to your online account to display your discounted pricing

Conditioning Reagent, For Sulfate, LabChem™

$89.60 - $89.60

Chemical Identifiers

Chemical Name or Material Conditioning Reagent
CAS 7732-18-5
CAS Max % 50.66
CAS 67-63-0
CAS Max % 20
CAS 7647-14-5
CAS Max % 15
CAS 56-81-5
CAS Max % 10
CAS 7647-01-0
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC133201

 
lab chem
LC133201
500mL Each for $89.60
Description & Specifications

Specifications

Formula Weight 58.44
Chemical Name or Material Conditioning Reagent
CAS Max % 15
Decomposition Information Hydrogen chloride; Carbon monoxide; Carbon dioxide
CAS 56-81-5
Color Clear, colorless
Quantity 500mL
CAS Max % 10
CAS 7647-01-0
CAS Max % 4.34
Name Note For Sulfate
Physical Form Liquid
Linear Formula NaCl
CAS 7732-18-5
UN Number UN1789
Health Hazard 1 Danger
CAS Max % 50.66
Health Hazard 2 GHS H Statement
Causes skin irritation.
Causes serious eye damage.
Identification Passes Test
CAS 67-63-0
Health Hazard 3 GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Packaging Poly Bottle
CAS Max % 20
Solubility Soluble in water
CAS 7647-14-5
Storage Information Room Temperature
Safety and Handling

Danger

GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical attention.
Take off contaminated clothing and wash before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.

GHS H Statement
Causes skin irritation.
Causes serious eye damage.