Please login to your online account to display your discounted pricing

Con-Trol 100, Osmometer Reference Standards, Precision Systems

$182.88 - $182.88

Chemical Identifiers

Chemical Name or Material CON-TROL 100
Physical Form Aqueous Solution
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

12-827-25A

 
precision systems inc
PSY2201
5mL/Ampoule Pack of 12 Each for $182.88
Description & Specifications

Specifications

Quantity 5mL/Ampoule
Physical Form Aqueous Solution
Chemical Name or Material CON-TROL 100

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.