Contec™ CONSTIX™ Surface Validation Swabs
Contec™ CONSTIX™ Surface Validation Swabs designed for the evaluation of surface cleaning protocols in biopharmaceutical industries.
$12.55 - $12.55
Specifications
Adhesive Type | Ultrasonically sealed |
---|---|
For Use With (Application) | Evaluation of surface cleaning protocols in bio-pharmaceutical industries |
Handle Material | Polypropylene |
Tip Material | Knit Polyester |
Product Type | Swab |
Catalog Number | Mfr. No. | Length (Metric) Tip | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) Tip | Price | Quantity | |||||
19-160-0021
|
Contec™
SV2 |
2 cm |
Case of 200 Each for $12.55
|
|
Description
- Package contains five polyester swabs specially processed to minimize background TOC contribution from the swab
- Low background TOC content for minimal interference with test results
- Large surface of the swab head and the long flexible handle provide exceptional surface coverage when swabbing
- Strong, durable polyester swabs for optimum retrieval
- Sterilizable by most common methods
- Autoclavable
- Swabs sealed in nylon pouch
Specifications
Ultrasonically sealed | |
Polypropylene | |
Swab |
Evaluation of surface cleaning protocols in bio-pharmaceutical industries | |
Knit Polyester |