Fisherbrand™ Complete Plumbing Assemblies for Lab Workstations
Fisherbrand™ Complete Plumbing Assemblies are furnished complete for Lab Workstations. Designed specifically for use in the laboratory environment.
Manufacturer:  Fisherbrand™ FEISVAL
 View more versions of this product
Catalog No. 06000356
Includes: Male Quick Connect, System Hose and Service Turret
Description
Description
  • Service turret is color coded base on service
  • Service turret is a 135° degreet turret with one polished chrome valve with a brass floating cone and integral brass seat.
  • Service turrets provide metering control of laboratory gases at pressures up to 60 PSI.
  • System service hoses carry services from the overhead service panel through the upright of the table and to the shank of the fixture. Hoses extend 48 in. above the workstation
  • System service hoses are color coded reinforced PVC
  • Male Quick connect: double end shut off plugs have an internal valve that shuts off flow when uncoupled
Specifications
Specifications
Color Coded Orange
Male quick connect, system hose and service turret
Reinforced PVC hose, polished chrome turret
Left Side, Air Service Complete Plumbing Assembly
Documents