Complete Optical Bench System
Quality, long-lasting instruments for use with a laser.
Manufacturer:  Fisher Scientific 45212EMD
 View more versions of this product
Catalog No. S42815ND
Requires :
  • Separate laser
Description
Description
  • Education-grade bench system designed with component interchangeability
  • Standard 1/4 in.-20 threads on mounting pins
  • Nearly universal exchangeability with other systems
Documents