Complete Optical Bench System
Quality, long-lasting instruments for use with a laser.
Manufacturer:  Fisher Scientific 45225EMD
 View more versions of this product
Catalog No. S42811ND
Requires :
  • Separate laser
Description
Description
  • Education-grade bench system designed with component interchangeability
  • Standard 1/4 in.-20 threads on mounting pins
  • Nearly universal exchangeability with other systems
Specifications
Specifications
Use with a laser
Two 23.6 in. benches, two leveling base sets, adjustable laser holder, heavy-duty pin carrier with pin, two ring mounts, two adjustable mirror holders, four standard pin carriers with pins
Optical bench system
Each
Documents