Please login to your online account to display your discounted pricing
Complete Cartesian Diver Kit
Cartesian diver demonstrates Archimedes' principle of buoyancy.
Manufacturer: fisher science education  GIG305
 View more versions of this product
Catalog No. S41121
  • / Each
Description & Specifications
Specifications
Height (English) 2.5 in.
Height (Metric) 6.35cm
For Use With (Application) Can be used as replacement to Cartesian Diver Unit or used in other cylinders or soda pop bottles experiments to demonstrates Archimedes' principle of buoyancy
Includes Blown Glass piece with a red band around the neck
Product Type Cartesian diver