missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Complete Blackboard Optics Kit

Revolutionary approach to the presentation of concepts in geometrical and physical optics.

Supplier:  Customer Channels Group (Ccg) K04100M

Catalog No. S42208-4


Includes:
  • Two ray projectors
  • Two project holders
  • Two 13 in. (32.5cm) clamping bars
  • 15 in. (37.5cm) Clamping bar
  • Plano convex lens
  • Plano concave lens
  • Semicircular lens
  • Parallel-sided block
  • Right-angle prism
  • Two plane mirrors
  • Double curved mirror
  • Projector lamp (spare)
  • Cushioned wooden storage case

Requires : Power supply (S422084A)

Description

Description

  • All aspects of the experiment can be done on the blackboard
  • All the constituent parts are secured to the blackboard by magnet or suction cup
  • Experiments, measurements, and calculations can be done without darkening the room
  • High visibility for demonstrations and lectures to large classes
  • Optical pieces are 2 × 8 in. (5 × 20cm)

Specifications

Specifications

Complete Blackboard Optics Kit
Revolutionary approach to the presentation of concepts in geometrical and physical optics
Magnetic grips, 2 Ray projectors, 2 project holders, 2 13 in. clamping bars, 15 in. clamping bar, plano convex lens, plano concave lens, semicircular lens, parallel-sided block, right angle prism, 2 plane mirrors, double curved mirror, projector lamp, cushioned wooden storage case
SDS
Documents

Documents

Promotions

Promotions