Please login to your online account to display your discounted pricing

CombiTitrant 2, AQUASTAR™, EMD Chemicals


Quantity 2.5L
Packaging GL45 Poly coated glass bottle

Manufacturer: emd millipore corporation  1880022545

 View more versions of this product

Catalog No. M1880022545

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Red-brown
Quantity 2.5L
Density 1.090-1.110g/cm3
Packaging GL45 Poly coated glass bottle

Specifications

Physical Form Liquid