Please login to your online account to display your discounted pricing

CombiSolvent Oils, AQUASTAR™, EMD Millipore


Quantity 1L

Manufacturer: emd millipore corporation  1880201045

 View more versions of this product

Catalog No. M1880201045

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 1L
Color Colorless to light yellow
Physical Form Liquid
Recommended Storage Room Temperature
Chemical Name or Material CombiSolvent Oils
Name Note AQUASTAR™, Karl Fischer Solvent for Mineral Oils

Specifications

Chemical Name or Material CombiSolvent Oils
Name Note AQUASTAR™, Karl Fischer Solvent for Mineral Oils
Physical Form Liquid
Accessories

Room Temperature