Please login to your online account to display your discounted pricing

CombiSolvent Oils, AQUASTAR™, EMD Millipore

$343.03 - $950.26

Chemical Identifiers

Chemical Name or Material CombiSolvent Oils
Name Note AQUASTAR™, Karl Fischer Solvent for Mineral Oils
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

M1880201045

 
emd millipore corporation
1880201045
1L Each for $950.26

M1880201000

 
emd millipore corporation
1880201000
1L Each for $343.03
Description & Specifications

Specifications

Quantity 1L
Color Colorless to light yellow
Physical Form Liquid
Chemical Name or Material CombiSolvent Oils
Recommended Storage Room Temperature
Name Note AQUASTAR™, Karl Fischer Solvent for Mineral Oils
Safety and Handling

Room Temperature