Please login to your online account to display your discounted pricing

CombiMethanol, 99.8% min, AQUASTAR™, EMD Millipore


Quantity 2.5L
Packaging GL45 Glass Bottle

Manufacturer: emd millipore corporation  1880092545

 View more versions of this product

Catalog No. M1880092545

  • / Each
 

Description & Specifications

Chemical Identifiers

Color Clear colorless
Physical Form Liquid
Formula Weight 32.04
Quantity 2.5L
Packaging GL45 Glass Bottle
Water 0.01% Max.
Solubility in Water To pass test
Substances Reducing Permanganate To pass test
Substances Darkened by H2SO4 To pass test
Residue after Evaporation 0.001% max
Chemical Name or Material CombiMethanol
Assay Percent Range 99.8% min.
CAS 67-56-1
Molecular Formula CH3OH
Titratable Base 0.0002meq/g max
Carbonyl Compounds 0.001% max
Titratable Acid 0.0003meq/g max

Specifications

Chemical Name or Material CombiMethanol
CAS 67-56-1
Physical Form Liquid
Assay Percent Range 99.8% min.
Molecular Formula CH3OH
Formula Weight 32.04