Please login to your online account to display your discounted pricing

CombiMethanol, 99.8% min, AQUASTAR™, EMD Millipore

$172.00 - $182.31

Chemical Identifiers

Chemical Name or Material CombiMethanol
CAS 67-56-1
Physical Form Liquid
Assay Percent Range 99.8% min.
Molecular Formula CH3OH
Formula Weight 32.04
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1880092545

 
emd millipore corporation
1880092545
GL45 Glass Bottle 2.5L Each for $182.31

M1880091045

 
emd millipore corporation
1.88009.1045
GL45 Glass Bottle 1L Each for $178.43

M1880091038

 
emd millipore corporation
1880091038
Glass Bottle 1L Each for $178.43

M1880092538

 
emd millipore corporation
1880092538
38/430 Glass Bottle 2.5L Each for $172.00
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Formula Weight 32.04
Quantity 2.5L
Packaging GL45 Glass Bottle
Water 0.01% Max.
Solubility in Water To pass test
Substances Reducing Permanganate To pass test
Substances Darkened by H2SO4 To pass test
Residue after Evaporation 0.001% max
Chemical Name or Material CombiMethanol
Assay Percent Range 99.8% min.
CAS 67-56-1
Molecular Formula CH3OH
Titratable Base 0.0002meq/g max
Carbonyl Compounds 0.001% max
Titratable Acid 0.0003meq/g max