Please login to your online account to display your discounted pricing

CombiMethanol, 99.8% min, AQUASTAR™, EMD Millipore


Quantity 1L
Packaging Glass Bottle

Manufacturer: emd millipore corporation  1880091038

 View more versions of this product

Catalog No. M1880091038

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 32.04
Color Clear colorless
Quantity 1L
Water 0.01% Max.
Substances Reducing Permanganate To pass test
Titratable Base 0.0002meq/g max
Residue after Evaporation 0.001% max
Chemical Name or Material CombiMethanol
Assay Percent Range 99.8% min.
Molecular Formula CH3OH
Solubility in Water To pass test
CAS 67-56-1
Carbonyl Compounds 0.001% max
Titratable Acid 0.0003meq/g max
Packaging Glass Bottle
Substances Darkened by H2SO4 To pass test

Specifications

Chemical Name or Material CombiMethanol
CAS 67-56-1
Physical Form Liquid
Assay Percent Range 99.8% min.
Molecular Formula CH3OH
Formula Weight 32.04