MilliporeSigma™ Supelco™ Column for Flash-Chromatography Assembly with Threaded Joints, Joint Size: ST/NS 45/40
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
$618.54 - $1011.40
Specifications
Column Type Flash Chromatography
For Use With (Application) Flash Chromatography
Joint Size ST/NS 45/40
View More Specs
Products 2
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Length (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Length (Metric) Price Quantity  
111008832
View Documents Promotion Details
MilliporeSigma™ Supelco™
Z2024791EA
1000 mL 57 mm 508 mm
Each for $618.54
111008587
View Documents Promotion Details
MilliporeSigma™ Supelco™
Z2024951EA
2000 mL 76 mm 560 mm
Each for $1,011.40
Description
Description
Specifications
Specifications
Flash Chromatography
Flash Chromatography
ST/NS 45/40
SDS
missing translation for 'documents'
missing translation for 'documents'