MilliporeSigma™ Supelco™ Column for Flash-Chromatography Assembly with Threaded Joints and Fritted Disc (Porosity C), Joint Size: ST/NS 29/32
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
$417.77 - $550.66
Specifications
Column Type Flash Chromatography
For Use With (Application) Flash Chromatography
Joint Size ST/NS 29/32
View More Specs
Products 4
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Length (Metric) Price Quantity  
Catalog Number Mfr. No. Capacity (Metric) Diameter (Metric) Outer Length (Metric) Price Quantity  
111009317
View Documents Promotion Details
MilliporeSigma™ Supelco™
Z4165761EA
100 mL 19 mm 813 mm
Each for $511.91
111009313
View Documents Promotion Details
MilliporeSigma™ Supelco™
Z4165841EA
200 mL 25 mm 584 mm
Each for $417.77
111009070
View Documents Promotion Details
MilliporeSigma™ Supelco™
Z4165921EA
400 mL 38 mm 560 mm
Each for $493.89
111008953
View Documents Promotion Details
MilliporeSigma™ Supelco™
Z4166061EA
600 mL 44 mm 572 mm
Each for $550.66
Description
Description
Specifications
Specifications
Flash Chromatography
Flash Chromatography
ST/NS 29/32
SDS
missing translation for 'documents'
missing translation for 'documents'