MilliporeSigma™ Supelco™ Column for Flash-Chromatography Assembly with Ball Joints, Standard, Joint Size: SJ 51/30
Flash chromatography is useful for rapid, preparative separations with moderate resolutions.
Supplier:  MilliporeSigma™ Supelco™ Z2024281EA
Catalog No. 111009059
Description
Description
Specifications
Specifications
560 mm
Flash Chromatography
400 mL
38 mm
Flash Chromatography
SJ 51/30