missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Colorful Periodic Chart

Shows essential element information clearly and colorfully.

$61.74 - $69.03

Specifications

Subject Science
Includes Element symbol, atomic number, element name, atomic mass and radioactivity
For Use With (Application) Shows essential element information clearly and colorfully
View More Specs

Products 2
Catalog Number Mfr. No. Height (Metric) Width (Metric) Product Type Price Quantity  
Catalog Number Mfr. No. Height (Metric) Width (Metric) Product Type Price Quantity  
S63055
View Documents Promotion Details
Customer Channels Group (Ccg)
S63055
96.5 cm 127 cm Periodic Chart Laminated Poster
Each for $61.74
 
S63056
View Documents Promotion Details
Customer Channels Group (Ccg)
S63056
21.6 cm 27.9 cm Periodic Chart Pad (x100 sheets)
Each for $69.03
 
Description

Description

  • Different colors highlight group similarities and reinforce different electron configurations
  • Contains the element symbol, atomic number, element name, atomic mass and radioactivity

Specifications

Specifications

Science
Element symbol, atomic number, element name, atomic mass and radioactivity
Shows essential element information clearly and colorfully
SDS
missing translation for 'documents'

missing translation for 'documents'