Please login to your online account to display your discounted pricing
ColorFlex™ Goggles
Colorful, sparkling goggles are fun and chemical-splash resistant.
Manufacturer: fisher science education  7306J
 View more versions of this product
 Alert
Straps contain some amount of latex.
Catalog No. S47618W
  • / Each
Description & Specifications
Specifications
Frame Color White
Material Polycarbonate
Type Safety Goggles
Frame Material PVC
Lens Tint Clear
Color White
Size Junior
Certifications/Compliance ANSI Z87.1-1989 standards
Surface Coating Antifog
Lens Coating Antifog