Please login to your online account to display your discounted pricing
ColorFlex™ Goggles
Colorful, sparkling goggles are fun and chemical-splash resistant.
Manufacturer: fisher scientific  7306
 View more versions of this product
 Alert
Straps contain some amount of latex.
Catalog No. S47617W
  • / Each
Description & Specifications
Specifications
Surface Coating Antifog
Lens Coating Antifog
Frame Color White
Material Polycarbonate
Lens Tint Clear
Certifications/Compliance ANSI Z87.1-1989 standards
Color White
Type Safety Goggles
Size Regular
Frame Material PVC