Please login to your online account to display your discounted pricing

Color Standard, Pt-Co, 50 Units APHA, Certified, LabChem™


Quantity 500mL
Packaging Amber Glass

Manufacturer: lab chem  LC132941

 View more versions of this product

Catalog No. LC132941

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Formula Weight 485.99
Color Clear, light orange
Absorbance Passes Test
CAS Max % 99.53
CAS 7647-01-0
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Traceability to NIST Traceable to NIST
CAS Max % 0.44
Density 1g/mL
CAS 16921-30-5
Chemical Name or Material Color Standard
Quantity 500mL
CAS Max % 0.02
CAS 7791-13-1
CAS Max % 0.01
Name Note Pt-Co, 50 Units APHA
Physical Form Liquid
Specific Gravity 1g/mL
Linear Formula K2(PtCl6)
CAS 7732-18-5
Decomposition Information Hydrogen chloride
Identification Passes Test
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Packaging Amber Glass
Solubility Soluble in water
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Color Standard
CAS 7732-18-5
CAS Max % 99.53
CAS 7647-01-0
CAS Max % 0.44
CAS 16921-30-5
CAS Max % 0.02
CAS 7791-13-1
CAS Max % 0.01
Name Note Pt-Co, 50 Units APHA
Purity Grade Certified Grade
Linear Formula K2(PtCl6)
Formula Weight 485.99
Accessories

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature