Please login to your online account to display your discounted pricing

Color Standard, Pt-Co, 20 Units APHA, Certified, LabChem™

$384.06 - $384.06

Chemical Identifiers

Chemical Name or Material Color Standard
CAS 7732-18-5
CAS Max % 99.821
CAS 7647-01-0
CAS Max % 0.17
CAS 16921-30-5
CAS Max % 0.005
CAS 7791-13-1
CAS Max % 0.004
Name Note Pt-Co, 20 Units APHA
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC132921

 
lab chem
LC132921
Amber Glass 500mL Each for $384.06
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 485.99
Chemical Name or Material Color Standard
Color Clear, light orange
Quantity 500mL
CAS Max % 0.005
Absorbance Passes Test
CAS 7791-13-1
CAS Max % 0.004
Name Note Pt-Co, 20 Units APHA
Physical Form Liquid
Specific Gravity 1g/mL
Linear Formula K2(PtCl6)
CAS 7732-18-5
Decomposition Information Hydrogen chloride
Identification Passes Test
CAS Max % 99.821
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 7647-01-0
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Amber Glass
Solubility Soluble in water
Traceability to NIST Traceable to NIST
CAS Max % 0.17
Recommended Storage Room Temperature
Density 1g/mL
CAS 16921-30-5
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature