missing translation for 'onlineSavingsMsg'
Learn More

Color Standard, Pt-Co, 10 Units APHA, Certified, LabChem™

Catalog No. LC132911
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC132911 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC132911 Supplier LabChem Supplier No. LC132911
Only null left

Specifications

Chemical Name or Material Color Standard
Name Note Pt-Co, 10 Units APHA
Packaging Amber Glass
CAS Max % 99.905, 0.09, 0.003, 0.002
CAS 16921-30-5, 7791-13-1, 7647-01-0, 7732-18-5
Identification Passes Test
Density 1g/mL
Physical Form Liquid
Linear Formula K2(PtCl6)
Color Orange
Solubility Soluble in water
Quantity 500 mL
Formula Weight 485.99
Grade Certified
Show More Show Less