missing translation for 'onlineSavingsMsg'
Learn More

Matching Fluid G, USP - for Color and Achromicity, Ricca Chemical

Supplier:  Ricca Chemical Company 4621G16

Catalog No. 46-21G-16


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Color Standard
109-52-4 , 7647-01-0 , 7758-99-8 , 10025-77-1 , 7791-13-1
Acidic
100°C
Miscible with water
Color Reference Standard
USP
Amber Glass Bottle
Yellow/Brown
0.0°C
Water Analyzer
Calibration Solution
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions