missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Color Standard, Color No. 3, Ricca Chemical

$605.62 - $605.62

Chemical Identifiers

CAS 16921-30-5,7647-01-0,7732-18-5,7791-13-1
Physical Form Liquid
Chemical Name or Material APHA / Hazen (Platinum-Cobalt)
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
223000316 SDS Ricca Chemical Company
RI2230.00316
500mL Each for $605.62
R2230003100 Ricca Chemical Company
RIR2230003100C
Glass amber 100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

16921-30-5,7647-01-0,7732-18-5,7791-13-1
Liquid
APHA / Hazen (Platinum-Cobalt)

Specifications

16921-30-5,7647-01-0,7732-18-5,7791-13-1
Clear light yellow to yellow brown
APHA / Hazen (Platinum-Cobalt)
Liquid
500mL