Please login to your online account to display your discounted pricing

Color Standard, APHA/Hazen (Platinum-Cobalt), Color = 500, Ricca Chemical


Quantity 500mL
Packaging Amber Glass

Manufacturer: ricca chemical company  RISC024080500C*

 View more versions of this product

Catalog No. SC024080500

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500mL
Purity Grade APHA/Hazen (Platinum-Cobalt)
CAS 7732-18-5
CAS Max % 0.09
CAS 7791-13-1
CAS Min % 0.11
CAS Max % 91.0
CAS Max % 10.74
CAS 16921-30-5
CAS Min % 0.09
CAS Max % 0.12
CAS 7647-01-0
CAS Min % 87.0
CAS Min % 10.32
Chemical Name or Material Color Standard
Packaging Amber Glass
Name Note Color= 500

Specifications

Chemical Name or Material Color Standard
Name Note Color= 500
CAS 7732-18-5
CAS Min % 87.0
CAS Max % 91.0
CAS 7647-01-0
CAS Min % 10.32
CAS Max % 10.74
CAS 16921-30-5
CAS Min % 0.11
CAS Max % 0.12
CAS 7791-13-1
CAS Min % 0.09
CAS Max % 0.09
Purity Grade APHA/Hazen (Platinum-Cobalt)